Learn More
Phospho-EEF2 (Thr56) Polyclonal Antibody, Invitrogen™
Descripción
This gene encodes a member of the GTP-binding translation elongation factor family. This protein is an essential factor for protein synthesis. It promotes the GTP-dependent translocation of the nascent protein chain from the A-site to the P-site of the ribosome. This protein is completely inactivated by EF-2 kinase phosporylation.
Especificaciones
Especificaciones
| Antígeno | Phospho-EEF2 (Thr56) |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | EEF2 |
| N.º de referencia del gen | P13639, P58252, P05197 |
| Alias de gen | EEF2, EF-2, EF2, Elongation factor 2 |
| Símbolos de los genes | EEF2 |
| Mostrar más |