missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Cortactin (Tyr421) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 10352953
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10352953 100 μL cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10352953 Proveedor Invitrogen™ N.º de proveedor 44854G

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Phospho-Cortactin (Tyr421) Polyclonal Antibody for Western Blot, ICC/IF

Cortactin (Cttn) is a ubiquitous actin-binding protein that was originally identified as a substrate for Src. It contributes to the organization of the actin cytoskeleton and cell shape. Cortactin also plays a role in the formation of lamellipodia and cell migration. It is accumulated in peripheral, actin-enriched structures of cells suggesting that cortactin facilitates actin network formation. Cortactin has four major domains of interest the N-terminal acidic (NTA) and tandem repeats domains and the C-terminal proline-rich and SH3 Domains. NTA associates with the Arp2/3 and WASP complex at F-actin branches. Cortactin is involved in promoting cell motility and invasion, including a critical role in invadopodia, actin rich-subcellular protrusions associated with degradation of the ECM by cancer cells. Cortactin is phosphorylated by src family kinases at Y421, Y466, and Y482 and S405 and S418 that are phosphorylated by Erk family kinases.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Phospho-Cortactin (Tyr421)
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Conjugado Unconjugated
Formulación Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
génica Cttn
N.º de referencia del gen Q14247, Q60598
Alias de gen 1110020L01Rik; amplaxin; cortactin; Cttn; Cttnb; EMS1; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); FLJ34459; mammary tumor and squamous cell carcinoma associated (p80/85 src substrate); oncogene EMS1; Oncogene SRC8; Src substrate cortactin; src8; Unknown (protein for MGC:137923)
Símbolos de los genes Cttn
Especie del huésped Rabbit
Inmunógeno The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of mouse cortactin that contains tyrosine 421.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13043, 2017
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.