Learn More
Phenylbutazone, 98%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar L03449.14
| Cantidad | 25g |
|---|
Ver más versiones de este producto
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| C19H20N2O2 | |
| MFCD00005500 | |
| phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion | |
| CHEBI:48574 | |
| CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Especificaciones
| Phenylbutazone | |
| Odorless | |
| MFCD00005500 | |
| UN2811 | |
| 14,7277 | |
| VYMDGNCVAMGZFE-UHFFFAOYSA-N | |
| 4-butyl-1,2-diphenylpyrazolidine-3,5-dione | |
| 4781 | |
| 308.38 |
| 50-33-9 | |
| C19H20N2O2 | |
| 25g | |
| 290080 | |
| phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion | |
| CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 308.38 | |
| CHEBI:48574 | |
| 98% |