Learn More
Phenanthroline, Tocris Bioscience™
Enhances hPSC differentiation into cranial placode cells
Marca: Tocris Bioscience 6385/100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phenanthroline is a enhances hPSC differentiation into cranial placode cells. Induces a 5-fold increase in cells expressing SIX1.Especificaciones
| 1,10-Phenanthroline | |
| 66-71-7 | |
| >99% | |
| 100 mg |