Learn More
MedChemExpress Phalloidin
Specifications
Specifications
| Cantidad | 1 mg |
| Requisitos de almacenamiento | Protect from light ∣ Powder -80°C 2 years, -20°C 1 year ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Condiciones de envío | Room temperature |
| CAS | 17466-45-4 |
| Peso molecular | 788.87 g/mol |
| Objetivo | NO Synthase ∣ MMP ∣ Endogenous Metabolite |