Learn More
Tocris Bioscience™ PH 797804
Descripción
PH 797804 is a potent and selective p38α/β inhibitor (IC50 values are 26 and 102 nM, respectively). Exhibits >100-fold selectivity for p38α/β over 71 other kinases, enzymes and GPCRs, including p38γ, p38δ, JNK and ERK2. Inhibits LPS-induced TNFα production in U937 monocytes in vitro. Suppresses acute and chronic inflammatory responses in rat models of arthritis. Orally active.
PH 797804 is the (-)-atropisomer, which is 100-fold more potent than its (+)-atropisomer.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (-)-3-[3-Bromo-4-[(2,4-difluorophenyl)methoxy]-6-methyl-2-oxo-1(2H)-pyridinyl]-N,4-dimethylbenzamide |
| CAS | 1358027-80-1 |
| Cantidad | 50 mg |
| Objetivo | p38 Inhibitors |
| Fórmula molecular | C22H19BrF2N2O3 |
| Formula Weight (peso de la fórmula) | 477.3 |
| Pureza | >98% |