Learn More
Tocris Bioscience™ PF 06273340, Tocris Bioscience™
Potent and selective pan-Trk inhibitor; peripherally restricted
Marca: Tocris Bioscience™ 6115/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PF 06273340 is a potent and selective pan-Trk inhibitor (IC50 values are 1, 2 and 6 nM for Trk C, B and A, respectively). Selective for Trk over a panel of ion channels, receptors and other enzymes. Peripherally restricted. Exhibits efficacy in a rodent model of inflammatory pain. Orally bioavailable.Especificaciones
| PF 06273340 | |
| N-[5-[[2-Amino-7-(2-hydroxy-1,1-dimethylethyl)-7H-pyrrolo[2,3-d]pyrimidin-5-yl]carbonyl]-3-pyridinyl]-5-chloro-2-pyridineacetamide | |
| Soluble to 100mM in DMSO | |
| Orange |
| 1402438-74-7 | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
Título del producto
For Research Use Only.