Learn More
Tocris Bioscience™ PF 06260933 dihydrochloride
MAP4K4 (HGK) inhibitor; also inhibits MINK and TNIK
Marca: Tocris Bioscience™ 5752/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PF 06260933 dihydrochloride is a MAP4K4 (HGK) inhibitor (IC50 = 140 nM). Also inhibits MINK and TNIK (IC50 values are 8 and 13 nM, respectively). Improves fasting hyperglycemia in mice. Orally active.Especificaciones
| 5-(4-Chlorophenyl)-[3,3 '-bipyridine]-6,6 '-diamine dihydrochloride | |
| 1883548-86-4 | |
| EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAP4K4, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, MEKKK4, mitogen-activa | |
| 10 mg |