Learn More
Tocris Bioscience™ PF 05190457
Descripción
PF 05190457 is a high affinity and selective ghrelin receptor inverse agonist (Kd = 3 nM). Exhibits selectivity for ghrelin receptors over a panel of other targets. Increases intracellular Ca2+ concentration in and insulin secretion from human islets in vitro. Increases gastric afferent nerve activity in a rat ex vivo stomach preparation.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[2-[(1R)-2,3-Dihydro-5-(6-methyl-4-pyrimidinyl)-1H-inden-1-yl]-2,7-diazaspiro[3,5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone |
| CAS | 1334782-79-4 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 25 mg |
| Fórmula molecular | C29H32N6OS |
| Formula Weight (peso de la fórmula) | Observed MW: 512.67 |