Learn More
PF 05180999, Tocris Bioscience™
GREENER_CHOICE
Potent and selective PDE2A inhibitor
Marca:  Tocris Bioscience 6405/5
Encompass
Código de producto. 15943281
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
PF 05180999 is a potent and selective PDE2A inhibitor (IC50 = 1 nM). Exhibits 2000-fold selectivity for PDE2A over PDE10A. Brain penetrant.
Especificaciones
Especificaciones
4-(1-Azetidinyl)-7-methyl-5-[1-methyl-5-[5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazol-4-yl]-imidazo[5,1-f][1,2,4]triazine
1394033-54-5
>98%