Learn More
PF 04671536 hydrochloride, Tocris Bioscience™
Potent and selective PDE8B/8A inhibitor
Marca: Tocris Bioscience 6024/25
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PF 04671536 hydrochloride is a potent and selective PDE8B/8A inhibitor ( IC50 values are 1.3 and 1.9 nM, respectively). Exhibits selectivity for PDE8A/B over other PDEs (IC50 values are >10 μM) and a range of other targets. Increases glucose-dependent insulin secretion from human pacreatic islet cells. Orally bioavailable.Especificaciones
| 5-Methyl-3-[[(2R)-4-(2-thiazolylmethyl)-2-morpholinyl]methyl]-3H-1,2,3-triazolo[4,5-d]pyrimidin-7-amine hydrochloride | |
| 1305116-67-9 | |
| >98% | |
| 25 mg |