Learn More
PF 04628935, Tocris Bioscience™
GREENER_CHOICE
Potent ghrelin receptor inverse agonist
Marca:  Tocris Bioscience 6347/25
Encompass
Código de producto. 15796409
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
PF 04628935 is a potent ghrelin receptor inverse agonist (IC50 = 4.6 nM). Orally bioavailable and exhibits reasonable brain penetration.
Especificaciones
Especificaciones
1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone
1383719-97-8
>98%