Learn More
Tocris Bioscience™ Perifosine
PKB/Akt inhibitor; also modulates Kv2.1 current
Marca: Tocris Bioscience™ 6087/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Perifosine is a PKB/Akt inhibitor; accumulates in and reorganizes lipid rafts disrupting Akt signaling. Also inhibits MAPK pathway and induces autophagy. Induces apoptosis in leukemia cell lines. Decreases Kv2.1 current amplitude by modulating its inactivation gating. Antitumor. Orally bioavailable.Especificaciones
| Perifosine | |
| Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate | |
| Soluble to 100mM in water and to 100mM in ethanol | |
| White |
| 157716-52-4 | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
Título del producto
For Research Use Only.