Learn More
Peretinoin, MedChemExpress
Código de producto. 16464196
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16464196 100 mg 100 miligramos
16435477 5 mg 5 miligramos
16419436 50 mg 50 miligramos
16412738 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16464196 Proveedor Medchem Express N.º de proveedor HY100008
MedChemExpress Peretinoin is an oral acyclic retinoid with a vitamin A-like structure that targets retinoid nuclear receptors such as retinoid X receptor (RXR) and retinoic acid receptor (RAR).
Peretinoin reduces the mRNA level of sphingosine kinase 1 (SPHK1) in vitro by downregulating a transcription factor, Sp1. Peretinoin prevents the progression of non-alcoholic steatohepatitis (NASH) and the development of hepatocellular carcinoma (HCC) through activating the autophagy pathway by increased Atg16L1 expression. Peretinoin inhibits HCV RNA amplification and virus release by altering lipid metabolism with a EC50 of 9 μM.
Especificaciones
Nombre del producto químico o material Peretinoin
CAS 81485-25-8
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C20H30O2
Cantidad 100 mg
Sinónimo NIK333
Información de solubilidad DMSO : 50 mg/mL (165.32 mM; Need ultrasonic)
SMILES C/C(C)=C/CC/C(C)=C/CC/C(C)=C/C=C/C(C)=C/C(O)=O
Peso molecular (g/mol) 302.45
Formula Weight (peso de la fórmula) 302.45
Porcentaje de pureza 95.66%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease