Learn More
Tocris Bioscience™ Pepstatin A Janelia Fluor 526
Descripción
Pepstatin A Janelia Fluor™ 526 is a fluorogenic green-emitting lysosome tracker and stain. No wash step is required when used for multicolor microscopy experiments. Pepstatin A Janelia Fluor™ 526 can be used for live cell imaging, in confocal microscopy and super-resolution microscopy techniques including 3D-SIM (structured illumination microscopy) and STED, as well as two-color lattice light-sheet microscopy (lls). Excitation maximum = 530 nm, emission maximum = 549 nm.
To measure the absorbance spectrum of this dye we recommend the following solvent: TFE plus 0.1% TFA.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-(3-(3,3-Difluoroazetidin-1-ium-1-ylidene)-6-(3,3-difluoroazetidin-1-yl)-2,7-difluoro-3 H-xanthen-9-yl)-4-(((6 S,9 S,12 S,13 S,17 S,20 S,21 S)-13,21-dihydroxy-12,20-diisobutyl-6,9-diisopropyl-2,17-dimethyl-4,7,10,15,18,23-hexaoxo-5,8,11,16,19,24-hexaazatriaconta |
| CAS | 2410614-07-0 |
| Cantidad | 100 μg |
| Objetivo | Fluorescent Lysosome Probes |
| Fórmula molecular | C67H91F6N9O12.CF3CO2H |
| Pureza | 0.9 |