Learn More
Penthiopyrad, MedChemExpress
Código de producto. 16447356
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16447356 100 mg 100 miligramos
16423407 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16447356 Proveedor Medchem Express N.º de proveedor HY17520
MedChemExpress Penthiopyrad(MTF-753) is a carboxamide fungicide used to control a broad spectrum of diseases on large variety of corps; inhibits fungal respiration by binding to mitochondrial respiratory complex II.
Penthiopyrad(MTF-753) is a carboxamide fungicide used to control a broad spectrum of diseases on large variety of corps
Especificaciones
Nombre del producto químico o material Penthiopyrad
Forma física Solid
CAS 183675-82-3
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C16H20F3N3OS
Sinónimo MTF-753
Información de solubilidad DMSO : ≥ 26 mg/mL (72.34 mM)
SMILES O=C(C1=CN(C)N=C1C(F)(F)F)NC2=C(C(C)CC(C)C)SC=C2
Peso molecular (g/mol) 359.41
Formula Weight (peso de la fórmula) 359.41
Porcentaje de pureza 97.45%