Learn More
Dibromuro de pentaeritritol-d8, TRC
Product Code. 30412885
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30412885 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412885 Supplier Toronto Research Chemicals Supplier No. TRCP26997210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Pentaerythritol-d8 Dibromide
CAS 1216413-92-1
Fórmula molecular C5H2D8Br2O2
Cantidad 10 mg
Sinónimo 2,2-Bis(bromomethyl)-1,3-propanediol-d8, 1,3-Dibromo-2,2-bis(hydroxymethyl) propane-d8, 1,3-Dibromo-2,2-dimethylolpropane-d8, Dibromoneopentyl-d8 Glycol, FR 1138-d8, FR 522-d8, NSC 9001-d8, Pentaerythritol-d8 Dibromohydrin
Fórmula InChI InChI=1S/C5H10Br2O2/c6-1-5(2-7,3-8)4-9/h8-9H,1-4H2/i1D2,2D2,3D2,4D2
SMILES [2H]C([2H])(O)C(C([2H])([2H])O)(C([2H])([2H])Br)C([2H])([2H])Br
Nombre IUPAC 2,2-bis[bromo(dideuterio)metil]-1,1,3,3-tetradeuteriopropano-1,3-diol
Peso molecular (g/mol) 269.99
Formula Weight (peso de la fórmula) 267.955