Learn More
Pemetrexed disodium hemipenta hydrate, MedChemExpress
Código de producto. 16466326
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
500 mg
Tamaño de la unidad:
100 miligramos
500 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16466326 100 mg 100 miligramos
16461706 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16466326 Proveedor Medchem Express N.º de proveedor HY13781
MedChemExpress Pemetrexed disodium hemipenta hydrate is a novel antifolate, the Ki values of the pentaglutamate of LY231514 are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.
Pemetrexed disodium hemipenta hydrate is a novel antifolate, the Ki values of the pentaglutamate of LY231514 are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.
Especificaciones
Nombre del producto químico o material Pemetrexed disodium hemipenta hydrate
CAS 357166-30-4
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H24N5Na2O8.5
Cantidad 100 mg
Sinónimo LY231514 disodium hemipenta hydrate
Información de solubilidad H2O : 100 mg/mL (193.64 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (3.87 mM; Need ultrasonic)
SMILES O=C(O[Na])CC[C@@H](C(O[Na])=O)NC(C1=CC=C(CCC2=CNC(N=C(N)N3)=C2C3=O)C=C1)=O.[2.5H2O]
Peso molecular (g/mol) 516.41
Formula Weight (peso de la fórmula) 516.41
Porcentaje de pureza 99.3%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death