Learn More
Thermo Scientific Chemicals Pectinasa, Aspergillus niger
Código de producto. p-7076612
Change view
Click to view available options
Cantidad:
250 mg
1 g
Tamaño de la unidad:
1 g
250 miligramos
15455659 250 mg 250 miligramos
15445659 1 g 1 g
2 options
Código de producto. 15455659
Marca: Thermo Scientific Chemicals J63408.MD
Solicitar formato a granel o personalizado
Edge
Se utiliza en la preparación de protoplastos vegetales para digerir la pared celular antes del aislamiento de los orgánulos. La pectinasa ayuda a facilitar la maceración de los materiales vegetales y los procesos de extracción, incluida la preparación de los orgánulos.
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 250 mg
Formulario Polvo