Learn More
Tocris Bioscience™ PEAQX tetrasodium salt
Descripción
PEAQX tetrasodium salt is a potent NMDA receptor antagonist (IC50 = 8 nM). Exhibits >100-fold selectivity for hGluN2A (formally hNR2A) containing receptors over hGluN2B (hNR2B). Anticonvulsant in vivo.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Especificaciones
Especificaciones
| Nombre del producto químico o material | [[[(1S)-1-(4-Bromophenyl)ethyl]amino](1,2,3,4-tetrahydro-2,3-dioxo-5-quinoxalinyl)methyl] phosphonic acid tetrasodium salt |
| CAS | 2102348-87-6 |
| Cantidad | 10 mg |
| Fórmula molecular | C17H13BrN3Na4O5P |
| Formula Weight (peso de la fórmula) | 542.14 |
| Pureza | 98% |