Learn More
PDE1-IN-2, MedChemExpress
Código de producto. 16483388
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
100 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16483388 10 mg 10 miligramos
16479916 50 mg 50 miligramos
16464626 100 mg 100 miligramos
16446117 5 mg 5 miligramos
16440769 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16483388 Proveedor Medchem Express N.º de proveedor HY101490
MedChemExpress PDE1-IN-2 is a PDE1 inhibitor extracted from patent WO2016/55618 A1, example 31. PDE1-IN-2 has IC50 values of 6 nM, 140 nM and 164 nM for PDE1C, PDE1B and PDE1A, respectvely. PDE1-IN-2 is developed for the research of neurodegenerative disorders and psychiatric disorders.
PDE1-IN-2 is a PDE1 inhibitor extracted from patent WO2016/55618 A1, example 31. PDE1-IN-2 has IC50 values of 6 nM, 140 nM and 164 nM for PDE1C, PDE1B and PDE1A, respectvely. PDE1-IN-2 is developed for the research of neurodegenerative disorders and psychiatric disorders.
Especificaciones
Nombre del producto químico o material PDE1-IN-2
Forma física Solid
CAS 1904611-63-7
Color Marrón
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C16H21BrN4O2
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Información de solubilidad DMSO : 25 mg/mL (65.57 mM; Need ultrasonic)
SMILES O=C1C2=NN=C(C3CCOCC3)N2C=C(Br)N1CC4CCCC4
Peso molecular (g/mol) 381.27
Formula Weight (peso de la fórmula) 381.27
Porcentaje de pureza 98.0%