Learn More
Tocris Bioscience™ PDD 00017273
Descripción
PDD 00017273 is a potent and selective poly (ADP ribose) glycohydrolase (PARG) inhibitor (IC50 = 26 nM). Exhibits >350-fold selectivity for PARG over a panel of ion channels, enzymes and receptors, including PARP1 and ARH3. Maintains PAR chains and induces DNA double-stranded breaks in cells following DNA damage. Decreases colony formation of ZR-75-1 BRCA1 WT cells and inhibits cancer cell survival. Cell permeable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-1,2,3,4-tetrahydro-N-(1-methylcyclopropyl)-3-[(2-methyl-5-thiazolyl)methyl]-2,4-dioxo-6-quinazolinesulfonamide |
| CAS | 1945950-21-9 |
| Cantidad | 1 mg |
| Objetivo | PARG Inhibitors |
| Fórmula molecular | C23H26N6O4S2 |
| Pureza | 98% |