Learn More
STEMCELL Technologies PD173074
Descripción
- PD173074 is a selective and potent, ATP-competitive inhibitor of FGFR
- It acts on both FGFR3 and FGFR1 (IC50 = 5 and 21.5 nM respectively), and also inhibits FGFR2, FGFR4 and KDR
- It is approximately 1000 times more potent than another common FGFR inhibitor SU5402
- (Koziczak et al.), Mohammadi et al.), Trudel et al.)
Especificaciones
Especificaciones
| CAS | 219580-11-7 |
| Cantidad | 10 mg |
| Fórmula molecular | C28H41N7O3 |
| Pureza | ≥98% |