Learn More
PD-L1/CD274 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B11D11, Proteintech
Código de producto. 16887945 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16887945 20 μL 20 microlitros
16877945 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16887945 Proveedor Proteintech N.º de proveedor 662481IG20UL
Mouse Monoclonal Antibody
PD-L1 (Programmed death receptor ligand 1, B7-H1, CD274) is a 40 kDa transmembrane protein and B7 family member. PD-L1 has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production, and PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion. Together with B7.1, PD-L1 can inactivate T cells and this relationship is a focal point of cancer therapeutic treatment. At least two isoforms of PDL-1 are known to exist.
Especificaciones
Antígeno PD-L1/CD274
Aplicaciones Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry, Western Blot
Clasificación Monoclonal
Clon 2B11D11
Concentración 1.55 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CD274
N.º de referencia del gen Q9EP73, Q9NZQ7
Alias de gen B7 H, B7 H1, B7 homolog 1, B7H1, CD274, CD274 molecule, PD L1, PDCD1 ligand 1, PDCD1L1, PDCD1LG1, PDL1, PD-L1, PD-L1/CD274, Programmed death ligand 1
Símbolos de los genes CD274
Especie del huésped Mouse
Inmunógeno PD-L1/CD274 Fusion Protein Ag12443
Método de purificación Protein G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 29126, 499342, 574058, 60533
Especies diana Human, Pig, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1