Learn More
Tocris Bioscience™ PD 98059
Descripción
PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.
Especificaciones
Especificaciones
| CAS | 167869-21-8 |
| Cantidad | 1 mg |
| Fórmula molecular | 2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one |
| Tipo de producto | PD 98059 |
| Pureza | 98% |
| Peso molecular | M.W. (Observed): 267.28 |
| Inhibidores | PD 98059 |
Para uso exclusivo en investigación