missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHB6 Monoclonal Antibody (2G11)

Descripción
Peptide Sequence: ASLRVRDIND HAPEFPAREM LLKISEITMP GKIFPLKMAH DLDTGSNGLQ RYTISSNPHF HVLTRNRSEG RKFPELVLDK PLDREEQPQL RLTLIALDGG.
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections.
Especificaciones
Especificaciones
| Antígeno | PCDHB6 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 2G11 |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | PCDHB6 |
| N.º de referencia del gen | Q9Y5E3 |
| Alias de gen | PCDHB6; PCDH-BETA6; PCDH-beta-6; POLR1E; protocadherin beta 6; protocadherin beta-6 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?