Learn More
DiscoverX PathHunter™ eXpress TAAR5 CHO-K1 β-Arrestin Orphan GPCR Assay
Descripción
The PathHunter™ TAAR5 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. methylamines) based activation of TAAR5 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged TAAR5 and EA-tagged β-Arrestin 2. Activation of TAAR5 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | GPCR |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | TAAR5 |
| Línea de células | CHO-K1 |
| Objetivo | TAAR5 |