Learn More
DiscoverX PathHunter™ eXpress mFPR2 CHO-K1 β-Arrestin GPCR Assay
Descripción
The PathHunter™ mFPR2 β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. formylpeptides) based activation of mFPR2 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged mFPR2 and EA-tagged β-Arrestin 2. Activation of mFPR2 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein.
Especificaciones
Especificaciones
| Entrada del ensayo | β-Arrestin-2 recruitment |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Agonista primario | WKYMVm-NH2 |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | mFPR2 |
| Línea de células | CHO-K1 |
| Objetivo | mFPR2 |