missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Park7/DJ-1 Mouse anti-Human, Mouse, Rat, Clone: 925805R, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB39951-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Knockout Validated.Especificaciones
| Park7/DJ-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DJ1, DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 11315 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry, KnockDown | |
| 925805R | |
| Western Blot 1 ug/mL, ELISA, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
| Q99497 | |
| PARK7 | |
| E. coli-derived recombinant human PARK-7 Ala2-Asp189 Accession # Q99497 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat PARK-7 in Western blots. In sandwich immunoassays, it is specific for human PARK-7. | |
| Human, Mouse, Rat | |
| Purified |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?