Learn More
Thermo Scientific Chemicals Parecoxib, sal sódica
Código de producto. 16524484 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16524484 1 g 1 g
16534484 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16524484 Proveedor Thermo Scientific Chemicals N.º de proveedor 465380010
Solicitar formato a granel o personalizado
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 198470-85-8
Clave InChI HQPVVKXJNZEAFW-UHFFFAOYSA-M
Nombre IUPAC sodio [4-(5-metil-3-fenil-1,2-oxazol-4-il)benzenosulfonil](propanoil)azanida
SMILES [Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1
Especificaciones
Nombre del producto químico o material Parecoxib sodium salt
Punto de fusión 273.0°C to 275.0°C
Fórmula molecular C19H17N2NaO4S
Cantidad 1 g
Peso molecular (g/mol) 392.40
Formula Weight (peso de la fórmula) 392.4