Learn More
Tocris Bioscience™ PAMP-20 (human)
Descripción
PAMP-20 (human) is an endogenous peptide agonist of Mas related GPR X2 (MRGPRX2, EC50 = 251 nM). Also a noncompetitive nicotinic cholinergic antagonist. Inhibits nicotine-stimulated catecholamine secretion from PC12 cells in vitro, and from sympathetic nerve endings and adrenal chromaffin cells in vivo. Corresponds to amino acids 1 - 20 of proadrenomedullin. Antihypertensive.
Especificaciones
Especificaciones
| Pureza | 95% |
| CAS | 150238-87-2 |
| Contenido y almacenamiento | Store at -20°C |
| Peso molecular | 2460.87 |
| Cantidad | 1 mg |