Learn More
Tocris™ Palonosetron hydrochloride
Descripción
Palonosetron hydrochloride is a highly potent and high affinity 5-HT3 antagonist (IC50 = 0.24 nM for 5-HT3A; pKi = 10.4). Displays a slow dissociation rate. Inhibits 5-HT-induced enhancement of cellular response to substance P (Cat. No. 1156) in vitro. Inhibits cisplatin (Cat. No. 2251)-induced enhancement of substance P response in vivo.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (3 AS)-2-[(3 S)-1-azabicyclo[2.2.2 ]oct-3-yl]-2,3,3 A,4,5,6-hexahydro-1 H-benz[de]isoquinolin-1-one hydrochloride |
| CAS | 135729-62-3 |
| Cantidad | 10 mg |
| Objetivo | 5-HT3 Receptor Antagonists |
| Fórmula molecular | C19H24N2O.HCl |
| Formula Weight (peso de la fórmula) | 808 |
| Pureza | 98% |