Learn More
Palbociclib, MedChemExpress
Product Code. 16477866
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
1 g
10 miligramos
100 miligramos
200 miligramos
5 miligramos
50 miligramos
500 miligramos
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16477866 100 mg 100 miligramos
16494017 50 mg 50 miligramos
16491316 200 mg 200 miligramos
16420568 5 mg 5 miligramos
16418108 10 mg 10 miligramos
16411926 500 mg 500 miligramos
16451939 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16477866 Supplier Medchem Express Supplier No. HY50767
This item is not returnable. View return policy
MedChemExpress Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
Specifications
Nombre del producto químico o material Palbociclib
CAS 571190-30-2
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C24H29N7O2
Cantidad 100 mg
Sinónimo PD 0332991
Información de solubilidad DMSO : 25 mg/mL (55.86 mM; ultrasonic and warming and adjust pH to 5 with HCl and heat to 60°C) ∣0.1 M HCL : 25 mg/mL (55.86 mM; ultrasonic and adjust pH to 4 with 0.1 M HCL) ∣H2O : 0.1 mg/mL (0.22 mM; Need ultrasonic)
SMILES O=C1C(C(C)=O)=C(C2=CN=C(N=C2N1C3CCCC3)NC4=CC=C(N5CCNCC5)C=N4)C
Peso molecular (g/mol) 447.53
Formula Weight (peso de la fórmula) 447.53
Porcentaje de pureza 95.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease