Learn More
Paeoniflorin, MedChemExpress
Código de producto. 16449046
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
200 mg
Tamaño de la unidad:
100 miligramos
200 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16449046 100 mg 100 miligramos
16401506 200 mg 200 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16449046 Proveedor Medchem Express N.º de proveedor HYN0293
MedChemExpress Paeoniflorin is a heat shock protein-inducing compound and commonly exists in the plants of Paeoniaceae family, with various biological activities, including anticancer activity, anti-inflammatory activity, enhancing cognition and attenuating learning impairment, anti-oxidative stress, antiplatelet aggregation, expansion of blood vessels, and reducing blood viscosity.
Paeoniflorin is a heat shock protein-inducing compound and commonly exists in the plants of Paeoniaceae family, with various biological activities, including anticancer activity, anti-inflammatory activity, enhancing cognition and attenuating learning impairment, anti-oxidative stress, antiplatelet aggregation, expansion of blood vessels, and reducing blood viscosity.
Especificaciones
Nombre del producto químico o material Paeoniflorin
CAS 23180-57-6
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C23H28O11
Cantidad 100 mg
Sinónimo Peoniflorin
Información de solubilidad DMSO : 100 mg/mL (208.13 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](O)[C@@H](O)[C@]([H])(O[C@]2([C@@]3(C)O[C@]4([H])O[C@@]5(C3)O)C[C@]5([H])[C@]24COC(C6=CC=CC=C6)=O)O[C@@H]1CO
Peso molecular (g/mol) 480.46
Formula Weight (peso de la fórmula) 480.46
Porcentaje de pureza 95.0%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neurodegeneration