Learn More
Tocris Bioscience™ PA Janelia Fluor™ 549, Haloalkane
Descripción
Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, super-resolution microscopy (SRM) techniques including PALM (in both live and fixed cells). Allows multicolor single tracking experiments.
Properties and Photophysical Data: PA Janelia Fluor™ 549, Haloalkane shows superior brightness and photostability.
Janelia Fluor™ is a registered trademark of Howard Hughes Medical Institute.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3',6'-Bis(1-azetidinyl)-N-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethyl]-2-diazo-2,3-dihydro-3-oxospiro[1H-indene-1,9'-[9H]xanthene]-6-carboxamide |
| CAS | 1811539-43-1 |
| Cantidad | 10 μg |
| Objetivo | Photoactivatable Dyes |
| Fórmula molecular | C38H42ClN5O5 |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | 90% |