Learn More
Tocris Bioscience™ P7C3
Descripción
P7C3 is a NAMPT activator. Proneurogenic and neuroprotective. Protects newborn neurons in the dentate gyrus by mitigating cell death. Also enhances learning and memory in aged rats. Orally available and brain penetrant. Also revents doxorubicin-mediated toxicity in osteosarcoma cells and restores intracellular NAD levels.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3,6-Dibromo-α-[(phenylamino)methyl]-9H-carbazole-9-ethanol |
| CAS | 301353-96-8 |
| Intervalo de porcentaje de ensayo | >99% |
| Cantidad | 25 mg |
| Fórmula molecular | C21H18Br2N2O |
| Formula Weight (peso de la fórmula) | Observed MW: 474.19 |