Learn More
Oxyresveratrol, MedChemExpress
Código de producto. 16462569
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
500 mg
1 g
Tamaño de la unidad:
1 g
100 miligramos
50 miligramos
500 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16462569 1 g 1 g
16454917 50 mg 50 miligramos
16442396 500 mg 500 miligramos
16419166 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16462569 Proveedor Medchem Express N.º de proveedor HYN1430
MedChemExpress Oxyresveratrol (trans-Oxyresveratrol) is a potent naturally occurring antioxidant and free radical scavenger (IC50 of 28.9 μM against DPPH free radicals). Oxyresveratrol is potent and noncompetitive tyrosinase inhibitor with an IC50 value of 1.2 μM for mushroom tyrosinase. Oxyresveratrol is effective against HSV-1, HSV-2 and varicella-zoster virus, and has neuroprotective effects.
Oxyresveratrol (trans-Oxyresveratrol) is a potent naturally occurring antioxidant and free radical scavenger (IC50 of 28.9 μM against DPPH free radicals). Oxyresveratrol is potent and noncompetitive tyrosinase inhibitor with an IC50 value of 1.2 μM for mushroom tyrosinase. Oxyresveratrol is effective against HSV-1, HSV-2 and varicella-zoster virus, and has neuroprotective effects.
Especificaciones
Nombre del producto químico o material Oxyresveratrol
CAS 29700-22-9
Color Earth Yellow
Notas de grado de pureza Research
Fórmula molecular C14H12O4
Cantidad 1 g
Sinónimo trans-Oxyresveratrol
Información de solubilidad DMSO : 50 mg/mL (204.72 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C(O)=C1
Peso molecular (g/mol) 244.24
Formula Weight (peso de la fórmula) 244.24
Porcentaje de pureza 98.87%
Grado Research
Forma física Solid