Learn More
Clorhidrato de oxifenciclimina, LoGiCal
Especificaciones
Especificaciones
| Nombre del producto químico o material | Oxyphencyclimine Hydrochloride |
| CAS | 125-52-0 |
| Nombres de analitos o componentes | Oxyphencyclimine Hydrochloride |
| Condiciones de envío | Room Temperature |
| Fórmula molecular | C20 H28 N2 O3 . Cl H |
| Certificaciones/conformidad | ISO 9001 |
| Sinónimo | Daricol, Daricon, Dominil, Enterex, NSC 528449, Oximin, Oxyphencyclimine hydrochloride, S 1-1236, Setrol, Syklifen, Ulcociclinina, Vio-Thene, Benzeneacetic acid, alpha-cyclohexyl-alpha-hydroxy-, (1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)methyl ester, hydrochloride (1:1), Benzeneacetic acid, alpha-cyclohexyl-alpha-hydroxy-, (1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)methyl ester, monohydrochloride (9CI), Cyclohexaneglycolic acid, alpha-phenyl-, (1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)methyl ester monohydrochloride (8CI), (1-Methyl-1,4,5,6-tetrahydro-2-pyrimidyl)methyl alpha-cyclohexyl-alpha-phenylglycolate hydrochloride, (+/-)-Oxyphencyclimine hydrochloride |
| Fórmula InChI | InChI=1S/C20H28N2O3.ClH/c1-22-14-8-13-21-18(22)15-25-19(23)20(24,16-9-4-2-5-10-16)17-11-6-3-7-12-17;/h2,4-5,9-10,17,24H,3,6-8,11-15H2,1H3;1H |
| Tipo de solución | Neat |
| SMILES | Cl.CN1CCCN=C1COC(=O)C(O)(C2CCCCC2)c3ccccc3 |
| Mostrar más |