Learn More
ORM-10103, MedChemExpress
Código de producto. 16478127
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16478127 5 mg 5 miligramos
16435418 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16478127 Proveedor Medchem Express N.º de proveedor HY128678
MedChemExpress ORM-10103 is a specific inhibitor of the Na+/Ca2+ exchanger (NCX), which decreases the NCX current with estimated IC50s of 55 and 67 nM at -80 and at 20 mV, respectively.
ORM-10103 is a specific inhibitor of the Na+/Ca2+ exchanger (NCX), which decreases the NCX current with estimated IC50s of 55 and 67 nM at -80 and at 20 mV, respectively.
Especificaciones
Nombre del producto químico o material ORM-10103
CAS 488847-28-5
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C20H16N2O4
Cantidad 5 mg
Información de solubilidad DMSO : 250 mg/mL (717.67 mM; Need ultrasonic)
SMILES O=[N+](C1=CN=C(OC2=CC=C3C(CCC(C4=CC=CC=C4)O3)=C2)C=C1)[O-]
Peso molecular (g/mol) 348.35
Formula Weight (peso de la fórmula) 348.35
Porcentaje de pureza 99.24%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation