Learn More
Oritavancin Phosphate, TRC
Código de producto. 30133265
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
25 mg
Tamaño de la unidad:
1 miligramo
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30133265 5 mg 5 miligramos
30132628 1 mg 1 miligramo
30009757 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30133265 Proveedor Toronto Research Chemicals N.º de proveedor TRCO6750505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 192564-14-0
Sinónimo (1S,2R,18R,19R,22S,25R,28R,40S)-2-[(2R,4S,5R,6S)-4-amino-5-hydroxy-4,6-dimethyloxan-2-yl]oxy-22-(2-amino-2-oxoethyl)-5,15-dichloro-48-[(2S,3R,4S,5S,6R)-3-[(2S,4S,5R,6S)-4-[[4-(4-chlorophenyl)phenyl]methylamino]-5-hydroxy-4,6-dimethyloxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-18,32,35,37-tetrahydroxy-19-[[(2R)-4-methyl-2-(methylamino)pentanoyl]amino]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentazaoctacyclo[26.14.2.23,6.214,17.18,12.129,33.010,25.034,39]pentaconta-3,5,8,10,12(48),14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid;phosphoric acid
Especificaciones
Formato Neat
Fórmula molecular C86H103Cl3N10O34P2
Peso molecular 1793.10298
Clave InChI PWTROOMOPLCZHB-BHYQHFGMSA-N
SMILES OP(O)(O)=O.OP(O)(O)=O.[H][C@]12NC(=O)[C@]([H])(NC(=O)[C@]3([H])NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](NC(=O)[C@@H](CC(C)C)NC)[C@H](O)C4=CC(Cl)=C(OC5=CC3=CC(OC3=C(Cl)C=C(C=C3)[C@H]1O[C@H]1C[C@](C)(N)[C@@H](O)[C@H](C)O1)=C5O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O[C@H]1C[C@](C)(NCC3=CC=C(C=C3)C3=CC=C(Cl)C=C3)[C@@H](O)[C@H](C)O1)C=C4)C1=CC(=C(O)C=C1)C1=C(C=C(O)C=C1O)[C@H](NC2=O)C(O)=O
Nombre IUPAC (1S,2R,18R,19R,22S,25R,28R,40S)-2-{[(2R,4S,5R,6S)-4-amino-5-hydroxy-4,6-dimethyloxan-2-yl]oxy}-22-(carbamoylmethyl)-5,15-dichloro-48-{[(2S,3R,4S,5S,6R)-3-{[(2S,4S,5R,6S)-4-[({4'-chloro-[1,1'-biphenyl]-4-yl}methyl)amino]-5-hydroxy-4,6-dimethyloxan-2-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-18,32,35,37-tetrahydroxy-19-[(2R)-4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34(39),35,37,46,49-pentadecaene-40-carboxylic acid; bis(phosphoric acid)
Cantidad 5 mg
Peso molecular (g/mol) 1989.10