Learn More
Oritavancin diphosphate, MedChemExpress
Código de producto. 16480376
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
2 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16480376 2 mg 2 miligramos
16474627 50 mg 50 miligramos
16468816 100 mg 100 miligramos
16411138 5 mg 5 miligramos
16408728 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16480376 Proveedor Medchem Express N.º de proveedor HYB1831A
MedChemExpress Oritavancin diphosphate (LY333328 diphosphate) is an orally active glycopeptide antibiotic with activity against gram-positive organisms. Oritavancin diphosphate shows antibacterial effect against multidrug-resistant S. pneumoniae. Oritavancin diphosphate inhibits cell wall synthesis and disrupts the membrane potential.
Oritavancin diphosphate (LY333328 diphosphate) is an orally active glycopeptide antibiotic with activity against gram-positive organisms. Oritavancin diphosphate shows antibacterial effect against multidrug-resistant S. pneumoniae. Oritavancin diphosphate inhibits cell wall synthesis and disrupts the membrane potential.
Especificaciones
Nombre del producto químico o material Oritavancin diphosphate
Forma física Solid
CAS 192564-14-0
Color Blanco
Cantidad 2 mg
Notas de grado de pureza Research
Fórmula molecular C86H103Cl3N10O34P2
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo LY333328 diphosphate
Información de solubilidad H2O : 50 mg/mL (25.14 mM; Need ultrasonic) ∣DMSO : 41.67 mg/mL (20.95 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](O[C@@](O[C@@H](C)[C@@H]2O)([H])C[C@]2(C)NCC3=CC=C(C4=CC=C(Cl)C=C4)C=C3)[C@@H](O[C@H](CO)[C@H]1O)OC5=C(OC6=CC=C([C@H]([C@H]7NC([C@H](NC)CC(C)C)=O)O)C=C6Cl)C=C([C@](NC([C@@H](NC7=O)CC(N)=O)=O)([H])C(N[C@](C8=O)([H])C9=CC%10=C(O)C=C9)=O)C=C5OC%11=CC=C([C@H]([C@](C(N[C@@](C(O)=O)([H])C%12=C%10C(O)=CC(O)=C%12)=O)([H])N8)O[C@@](O[C@@H](C)[C@@H]%13O)([H])C[C@@]%13(N)C)C=C%11Cl.O=P(O)(O)O.O=P(O)(O)O
Peso molecular (g/mol) 1989.09
Formula Weight (peso de la fórmula) 1989.09
Porcentaje de pureza 99.66%