Learn More
Mezcla de plaguicidas organofosforados 580 400 μg/ml en acetona:hexano, Dr. Ehrenstorfer
Especificaciones
Especificaciones
| Nombre del producto químico o material | (EZ)-Tetrachlorvinphos∣Anilazine∣Atrazine∣Azinphos-methyl∣Chlorpyrifos∣Coumaphos∣Demeton (O+S)∣Diazinon∣Dichlorvos∣Dimethoate∣Disulfoton∣EPN∣Ethoprophos∣Famphur∣Fenchlorphos∣Fensulfothion∣Fenthion∣Malathion∣Merphos∣Mevinphos∣Monocrotophos∣Naled∣O,O,O-Triethylphosphorothioate∣O,O-TEPP∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Propazine∣Prothiophos∣Simazine∣Sulfotep∣Sulprofos∣Thionazin∣Trichloronate |
| CAS | 961-11-5, 101-05-3, 1912-24-9, 86-50-0, 2921-88-2, 56-72-4 |
| Nombres de analitos o componentes | (EZ)-Tetrachlorvinphos∣Anilazine∣Atrazine∣Azinphos-methyl∣Chlorpyrifos∣Coumaphos∣Demeton (O+S)∣Diazinon∣Dichlorvos∣Dimethoate∣Disulfoton∣EPN∣Ethoprophos∣Famphur∣Fenchlorphos∣Fensulfothion∣Fenthion∣Malathion∣Merphos∣Mevinphos∣Monocrotophos∣Naled∣O,O,O-Triethylphosphorothioate∣O,O-TEPP∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Propazine∣Prothiophos∣Simazine∣Sulfotep∣Sulprofos∣Thionazin∣Trichloronate |
| Condiciones de envío | +20°C |
| Certificaciones/conformidad | ISO 17034 |
| Solvent or Matrix | Acetone/Hexane |
| Cantidad | 1 mL |
| Concentration or Composition (by Analyte or Components) | (EZ)-Tetrachlorvinphos 400 μg/mL∣Anilazine 400 μg/mL∣Atrazine 400 μg/mL∣Azinphos-methyl 400 μg/mL∣Chlorpyrifos 400 μg/mL∣Coumaphos 400 μg/mL∣Demeton (O+S) 400 μg/mL∣Diazinon 400 μg/mL∣Dichlorvos 400 μg/mL∣Dimethoate 400 μg/mL∣Disulfoton 400 μg/mL∣EPN 400 μg/mL∣Ethoprophos 400 μg/mL∣Famphur 400 μg/mL∣Fenchlorphos 400 μg/mL∣Fensulfothion 400 μg/mL∣Fenthion 400 μg/mL∣Malathion 400 μg/mL∣Merphos 400 μg/mL∣Mevinphos 400 μg/mL∣Monocrotophos 400 μg/mL∣Naled 400 μg/mL∣O,O,O-Triethylphosphorothioate 400 μg/mL∣O,O-TEPP 400 μg/mL∣Parathion-ethyl 400 μg/mL∣Parathion-methyl 400 μg/mL∣Phorate 400 μg/mL∣Propazine 400 μg/mL∣Prothiophos 400 μg/mL∣Simazine 400 μg/mL∣Sulfotep 400 μg/mL∣Sulprofos 400 μg/mL∣Thionazin 400 μg/mL∣Trichloronate 400 μg/mL |