missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2A5/OR2A14 Polyclonal Antibody

Description
Antibody detects endogenous levels of total OR2A5/2A14.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
| Antígeno | OR2A5/OR2A14 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | OR2A14 |
| N.º de referencia del gen | Q96R47, Q96R48 |
| Alias de gen | olfactory receptor 2A14; Olfactory receptor 2A26; olfactory receptor 2A5; Olfactory receptor 2A6; olfactory receptor 2A8; Olfactory receptor 7-138/7-141; olfactory receptor family 2 subfamily A member 14; olfactory receptor family 2 subfamily A member 5; olfactory receptor OR7-12; olfactory receptor, family 2, subfamily A, member 11 pseudogene; olfactory receptor, family 2, subfamily A, member 14 pseudogene; olfactory receptor, family 2, subfamily A, member 26; olfactory receptor, family 2, subfamily A, member 5; olfactory receptor, family 2, subfamily A, member 6; olfactory receptor, family 2, subfamily A, member 8; OR2A11P; OR2A14; OR2A14P; OR2A26; OR2A5; OR2A6; OR2A8; OR7-138; OR7-141; OST182 |
| Símbolos de los genes | OR2A14, OR2A5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?