missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR10C1 Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total OR10C1.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Especificaciones
Especificaciones
| Antígeno | OR10C1 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | OR10C1 |
| N.º de referencia del gen | Q96KK4 |
| Alias de gen | hs6M1-17; Olfactory receptor 10C1; olfactory receptor 10C2; olfactory receptor family 10 subfamily C member 1 (gene/pseudogene); olfactory receptor OR6-31; olfactory receptor, family 10, subfamily C, member 1; olfactory receptor, family 10, subfamily C, member 2; OR10C1; OR10C1P; OR10C2; OR6-31; seven transmembrane helix receptor |
| Símbolos de los genes | OR10C1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?