Learn More
B Braun™ Omnican™ 20 U Insulin Injection Syringe
Descripción
- Single-use
- Integrated needle
- Latex-free, PVC-free, and DEHP-free
- Piston stop to prevent slipping out of the barrel
- Three-facet needle tip grinding
- Thin-wall needle (acc. to EN ISO 9626)
- Sterilized by ethylene oxide
Especificaciones
Especificaciones
| Certificaciones/conformidad | EN ISO 8537, MDD 93/42/EEC, class Iia, EN ISO 13485, EN ISO 9626 |
| Material | IR, Polyethylene, Polypropylene, Polystyrene, Silicone, Stainless Steel (Tip) |
| Calibre de aguja | 30 G |
| Graduaciones | 0.5 I.U. |
| Envase | 100 x 1 Pcs. |
| Volumen (métrico) | 0.5 mL |
| Graduated | Sí |
| Esterilidad | EO Sterile |