missing translation for 'onlineSavingsMsg'
Learn More

Okadaic acid sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 15464839
Change view
Click to view available options
Quantity:
100 μg
300 μg
50 μg
Unit Size:
100µg
300µg
50µg
Product Code. Quantity unitSize
15464839 50 μg 50µg
15444839 100 μg 100µg
15454839 300 μg 300µg
3 options
This item is not returnable. View return policy

Product Code. 15464839

Brand: Thermo Scientific Alfa Aesar J62463.I&

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A)
  • Does not inhibit tyrosine or alkaline phosphatases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209266-80-8
Molecular Formula C44H67NaO13
Molecular Weight (g/mol) 826.997
MDL Number MFCD00210208
InChI Key BYHIFOCTDVNQQT-GHIYGBLASA-M
Synonym okadaic acid sodium salt, okadaic acid sodium salt from prorocentrum concavum, okadaic acid sodium salt from prorocentrum concavum hplc , film
PubChem CID 71308608
IUPAC Name sodium;(2R)-3-[(2S,6R,8S,11R)-2-[(E,2R)-4-[(2S,2'R,4R,4aS,6R,8aR)-4-hydroxy-2-[(1S,3S)-1-hydroxy-3-[(2S,3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]
SMILES CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[Na+]

Specifications

Color White
Physical Form Powder
Quantity 50 μg
UN Number UN3462
Solubility Information Soluble in DMSO or ethanol
Formula Weight 827
Percent Purity 98%
Chemical Name or Material Okadaic acid sodium salt
compliance-icons
Product Identifier
  • Okadaic acid sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : Okadaic acid sodiumsalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.