Learn More
OICR-9429, MedChemExpress
Código de producto. 16457248
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16457248 10 mg 10 miligramos
16447216 100 mg 100 miligramos
16429777 5 mg 5 miligramos
16423566 25 mg 25 miligramos
16423267 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16457248 Proveedor Medchem Express N.º de proveedor HY16993
MedChemExpress OICR-9429 is high affinity WD repeat domain 5 (WDR5) inhibitor, competitively blocks WDR5 interaction with MLL protein via binding the central peptide-binding pocket of WDR5. OICR-9429 can suppress histone H3K4 trimethylation and can be used for the research of various cancers including non-MLL-rearranged leukaemia, colon, pancreatic, prostate cancer and bladder cancer (BCa) .
OICR-9429 is high affinity WD repeat domain 5 (WDR5) inhibitor, competitively blocks WDR5 interaction with MLL protein via binding the central peptide-binding pocket of WDR5. OICR-9429 can suppress histone H3K4 trimethylation and can be used for the research of various cancers including non-MLL-rearranged leukaemia, colon, pancreatic, prostate cancer and bladder cancer (BCa) .
Especificaciones
Nombre del producto químico o material OICR-9429
Forma física Solid
CAS 1801787-56-3
Color Blanco
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C29H32F3N5O3
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : ≥ 32 mg/mL (57.60 mM)
SMILES O=C(C1=CNC(C=C1C(F)(F)F)=O)NC2=CC(C3=CC(CN4CCOCC4)=CC=C3)=CC=C2N5CCN(C)CC5
Peso molecular (g/mol) 555.59
Formula Weight (peso de la fórmula) 555.59
Porcentaje de pureza 98.1%