Learn More
Tocris Bioscience™ OF 1
Selective BRPF1B and BRPF2 inhibitor
Marca: Tocris Bioscience™ 5289/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
OF 1 is a selective BRPF1B and BRPF2 bromodomain inhibitor (Kd values are 100 and 500 nM respectively). Exhibits 39-fold selectivity for BRPF1B and BRPF2 over BRD4. Accelerates FRAP recovery at 5 μM in a BRPF2 FRAP assay. Inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.Especificaciones
| 98% | |
| 919973-83-4 | |
| 10 mg |
| 4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide | |
| 4°C |
Título del producto
For Research Use Only