Learn More
Tocris Bioscience™ OF 1
GREENER_CHOICE
Selective BRPF1B and BRPF2 inhibitor
Marca:  Tocris Bioscience™ 5289/50
Encompass
Código de producto. 15766666
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
OF 1 is a selective BRPF1B and BRPF2 bromodomain inhibitor (Kd values are 100 and 500 nM respectively). Exhibits 39-fold selectivity for BRPF1B and BRPF2 over BRD4. Accelerates FRAP recovery at 5 μM in a BRPF2 FRAP assay. Inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.
Especificaciones
Especificaciones
98%
919973-83-4
50 mg
4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide
4°C
Certificados
Promociones
Promociones
Título del producto
Seleccione un problema
For Research Use Only