Learn More
Octenidine Dihydrochloride, TRC
Código de producto. 17922835
Change view
Click to view available options
Cantidad:
1 g
5 g
25 g
Tamaño de la unidad:
1 g
25 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
17922835 1 g 1 g
17973805 25 g 25 g
17910345 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 17922835 Proveedor Toronto Research Chemicals N.º de proveedor TRCO2391501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 70775-75-6
Fórmula molecular C36 H62 N4 . 2 H Cl
Peso molecular (g/mol) 623.83
Sinónimo N,N′-(1,10-decanediyldi-1(4H)-pyridinyl-4-ylidene)bis-1-Octanamine Dihydrochloride, Neo Kodan, Octeniderm, Octenidine 2HCl;Octenidine Dihydrochloride, Octenidine Hydrochloride, Octenidol, Octenilin, Win 41464-2
Nombre IUPAC N-octyl-1-[10-[4-(octylamino)pyridin-1-ium-1-yl]decyl]pyridin-1-ium-4-amine;dichloride
SMILES CCCCCCCC/N=C(C=C1)/C=CN1CCCCCCCCCCN(C=C/2)C=CC2=N/CCCCCCCC.[2ClH]
Especificaciones
Nombre del producto químico o material Octenidine Dihydrochloride
Notas de grado de pureza HPLC
Cantidad 1 g
Fórmula InChI InChI=1S/C36H62N4.2ClH/c1-3-5-7-9-15-19-27-37-35-23-31-39(32-24-35)29-21-17-13-11-12-14-18-22-30-40-33-25-36(26-34-40)38-28-20-16-10-8-6-4-2;;/h23-26,31-34H,3-22,27-30H2,1-2H3;2*1H
Formula Weight (peso de la fórmula) 622.4508
Porcentaje de pureza >95%